Learn More
ATCC Microsporum gypseum (Bodin) Guiart et Grigorakis (ATCC 14683)
Supplier:  ATCC 14683
Encompass
Microsporum gypseum (Bodin) Guiart et Grigorakis (ATCC 14683) is a dermatophyte fungal strain supplied as a freeze-dried ampoule containing viable cells for research, susceptibility testing, and reference use. It is classified as Biosafety Level 2; follow appropriate biosafety procedures when reviving and culturing this organism.
  • Product format: Freeze-dried ampoule containing viable cells.
  • Biosafety level: 2.
  • Recommended medium: Emmons' modification of Sabouraud's agar/broth.
  • Typical incubation temperature: 26 °C.
  • Storage recommendation: Store in the vapor phase of liquid nitrogen for long-term storage.
  • Strain designations and synonyms: PCI M-82 [IFO 8231]; synonym Arthroderma gypseum.
Catalog No. NC3893096