Learn More
MilliporeSigma™ Supelco™ MicroTee
Catalog No. 111010251 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
1/16 in. O.D. Tubing
1/32 in. O.D. Tubing
360 μm O.D. Tubing
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
11-101-0251 1/16 in. O.D. Tubing
11-101-0249 1/32 in. O.D. Tubing
11-101-0250 360 μm O.D. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-0251 Supplier MilliporeSigma™ Supelco™ Supplier No. 51286U
Add to Cart
Add to Cart
Connects 3 capillary tubes, using MicroFingertight II fittings. Includes 3 MicroFerrules for 0.025 in. O.D. tubing, 3 female nuts.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
Includes 3, microferrules for 1/16 in. O.D. tubing and 3, female nuts (5/16 in. -24 internal threads)
For Use With 1/16 in. O.D. Tubing