Learn More
MilliporeSigma™ Supelco™ MicroTight Adapters PEEK
Catalog No. 111011511 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With:
1/16 in. to 1/32 in. Tubing
Catalog No. For Use With
11-101-1511 1/16 in. to 1/32 in. Tubing
1 options
Catalog No. 11-101-1511 Supplier MilliporeSigma™ Supelco™ Supplier No. 51272U
Add to Cart
Add to Cart
These adapters will help create a true zero dead volume (ZDV) connection between 1/16 in. I.D. tubing and capillary tubing. MicroTight Adapters allow connection of 1/16 in. O.D. tubing directly to 1/32 in. O.D. and 360 μm tubing.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 1/Pk.
For Use With 1/16 in. to 1/32 in. Tubing
Material PEEK