Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Microtiter Tube Plug Strip
GREENER_CHOICE

Catalog No. 21403159 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Sterile
Quantity:
120/Pk., 5/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Quantity
21-403-159 Sterile 120/Pk., 5/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-403-159 Supplier Thermo Scientific™ Supplier No. 342511
  • $98.30
    $70.65 / Pack of 120

    Save $27.65

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $413.00
    $296.65 / Case of 5 PK

    Save $116.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.

Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.

  • LDPE non-autoclavable
  • Eight-plug strips can be easily cut apart for individual samples
  • Available sterile and nonsterile

TRUSTED_SUSTAINABILITY

Specifications

Product Type 8-cap Strip
Autoclavable Not Autoclavable
Description 1.2mL Microtiter Tube 8-cap plug strip
Material LDPE
Color Natural
Packaging 10 sealed bags of twelve 8 cap strips/unit, 5 units/case
Quantity 120/Pk., 5/Cs.
Sterility Sterile
For Use With (Equipment) Plug Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.