Learn More
Thermo Scientific™ Microtiter Tubes and Racks
GREENER_CHOICE
Catalog No. 21403208
Change view
Click to view available options
Product Type:
Microtiter Tube
Sterility:
Non-sterile
Sterile
Format:
Bulk
Robotic Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Sterility Format
21-403-208 Microtiter Tube Sterile Robotic Rack
21-403-212 Microtiter Tube Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-208 Supplier Thermo Scientific™ Supplier No. 3487
Add to Cart
Add to Cart
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Color Natural
Volume (Metric) 1.2 mL
Quantity 96 tubes/rack 10 racks/unit, 5 units case
Closure Material Polyethylene (Plug)
Product Line Thermo Scientific
Sterility Sterile
Format Robotic Rack
Product Type Microtiter Tube