Learn More
Invitrogen™ MICS1 Monoclonal Antibody (42), Invitrogen™
Description
Specificity: Dermatophagoides pteronyssinus GHITM/MICS1
Specifications
Specifications
| Antigen | MICS1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 42 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281 |
| Host Species | Mouse |
| Immunogen | Recombinant Dermatophagoides pteronyssinus GHITM/MICS1 Protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.