Learn More
Integra™ Miltex™ Mid Grade Operating Scissors
Catalog No. 12460784
Change view
Click to view available options
Length (Metric):
11.4 cm
12.7 cm
14 cm
16.5 cm
19 cm
Blade Style:
Curved
Straight
Tip Style:
Blunt/Blunt
Sharp/Blunt
Sharp/Sharp
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade Style Tip Style
12-460-784 14 cm Straight Blunt/Blunt
12-460-794 14 cm Curved Blunt/Blunt
12-460-791 16.5 cm Curved Sharp/Sharp
12-460-783 11.4 cm Straight Blunt/Blunt
12-460-792 12.7 cm Straight Sharp/Sharp
12-460-796 14 cm Straight Sharp/Sharp
12-460-797 16.5 cm Straight Sharp/Sharp
12-460-785 16.5 cm Straight Blunt/Blunt
12-460-778 14 cm Straight Sharp/Blunt
12-460-773 11.4 cm Straight Sharp/Blunt
12-460-781 19 cm Straight Sharp/Blunt
12-460-790 11.4 cm Curved Sharp/Sharp
12-460-782 11.4 cm Straight Sharp/Sharp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 12-460-784 Supplier Integra™ Miltex™ Supplier No. V9526
Add to Cart
Add to Cart
For general utility applications
  • Standard pattern
  • Blunt-blunt and sharp-blunt tips are ideal for cutting sutures, gauze, tubing and other materials
  • Sharp-sharp tips are preferred for cutting tendons and cartilage
  • Mid-grade forged from Japanese stainless steel
  • Properly hardened, tempered and passivated
  • Electrolytically polished and ultrasonically cleaned for a high quality finish
  • Guaranteed to be free from defects in workmanship and material
Specifications
Blade Style Straight
Length (English) 5.5 in.
Length (Metric) 14 cm
For Use With (Application) Surgical, Precision
Tip Style Blunt/Blunt