Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K1 Series Mid-Sole Ice Cleat

The K1 Series Mid-Sole ice cleat is the one-size-fits-all winter traction solution. The ice-penetrating tungsten carbide spikes maximize grip while the rotatable design provides seamless transition when traction is not required.

$39.75 - $40.70

Specifications

Closure Type Elastic
Color Black
For Use With (Application) Outdoor Surfaces, Ice Surfaces
Size Universal
Strap Material Elastic
View More Specs

Products 3
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
19-060-834
View Documents
K1 Series
V9770470O/S
Original Profile Tungston Carbide Molded in
Pair for $40.70
Only null left
Add to Cart
 
19-060-835
View Documents
K1 Series
V9770570O/S
Low Profile Tungston Carbide Molded in
Pair for $39.75
Only null left
Add to Cart
 
19-060-837
View Documents
K1 Series
V9770770O/S
High Profile Tungston Carbide Molded in
Pair for $40.70
Only null left
Add to Cart
 
Description

Description

  • Available in Original, Low-Profile, and High-Profile styles to provide near-universal coverage of all footwear
  • Strategically placed ice-penetrating tungsten carbide spikes provide maximum traction on ice and snow
  • Easy to use design rotates to top of foot to allow driving, climbing, and indoor use
  • Black or hi-viz adjustable strap delivers a perfect fit regardless of shoe size
  • Specially formulated rubber pad provides maximum comfort and durability even in extreme cold weather conditions
Specifications

Specifications

Elastic
Outdoor Surfaces, Ice Surfaces
Elastic
Unisex
Black
Universal
Reusable
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.