Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Middlebrook 7H11 Agar
GSA_VA

Catalog No. R01603
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
1 oz. Bottle
15mm x 60mm Plate
20mm x 113mm Tube
Monoplate Deep Pour
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01603 10/Pk. 15mm x 60mm Plate
R01605 10/Pk. Monoplate Deep Pour
R08570 100/Pk. 20mm x 113mm Tube
R10110 100/Pk. 1 oz. Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R01603 Supplier Thermo Scientific™ Supplier No. R01603
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and cultivate Mycobacterium species with this selective, ready-to-use medium.

Isolate and cultivate Mycobacterium species with Thermo Scientific™ Remel™ Middlebrook 7H11 Agar. Middlebrook 7H11 Agar is a modification of 7H9 and 7H10 media by Middlebrook. Casein hydrolysate was incorporated into this modified medium to stimulate the growth of mycobacteria that would not otherwise grow on this medium. Malachite-green dye is incorporated as a selective agent which inhibits bacteria other than mycobacteria.

Mycobacteria are immobile, slow-growing, rod-shaped, gram-positive bacteria with high genomic content. The genus includes pathogens known to cause serious diseases in mammals, including tuberculosis and leprosy.

Use Middlebrook 7H11 Agar for the isolation and cultivation of Mycobacterium species.

  • Selective: Malachite-green dye is incorporated in the medium acts as a selective agent which inhibits bacteria other than mycobacteria.
  • Ready-to-use: convenience of prepared media.

Middlebrook 7H11 Agar contains inorganic salts which are essential for the growth of mycobacteria. Casein hydrolysate serves as a growth stimulant for drug-resistant strains of Mycobacterium tuberculosis. Glycerol is a source of carbon and energy. Sodium citrate is converted to citric acid, which holds inorganic cations in solution.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Middlebrook 7H11 Agar
Format 15mm x 60mm Plate
Product Type Agar Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.