Learn More
Liofilchem™ Middlebrook 7H11 Agar
SDP
Catalog No. 21403469 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
Catalog No. Quantity
21-403-469 20 Plates
1 options
Catalog No. 21-403-469 Supplier Liofilchem™ Supplier No. 10416
Add to Cart
Add to Cart
A selective agar based medium for the isolation and cultivation of mycobacteria
  • Middlebrook 7H11 Agar is a specialized, agar-based culture medium for the selective isolation, cultivation, and susceptibility testing of Mycobacterium spp., including Mycobacterium tuberculosis, from clinical samples, especially those with mixed bacteria.
  • It is designed to grow fastidious mycobacteria, offering advantages like better drug retention for testing compared to older egg-based media, and facilitating rapid detection through microcolony methods.
  • This agar-based medium provides improved susceptibility testing compared to media based on coagulated egg (Lowenstein formulations) where drugs may be more easily inactivated.
Specifications
Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Mycobacteria
Test Type Microbial Cultivation
Quantity 20 Plates
Diameter (Metric) 90 mm