Learn More
Ace Glass, Inc. Midget Impinger, Complete with 0-30mL (5mL divisions)graduated bottle and #7 Ace-Safe™; stopper with nozzle tip
Supplier: Ace Glass, Inc. 753323
Midget Impinger, Complete with 0-30mL (5mL divisions)graduated bottle and #7 Ace-Safe™; stopper with nozzle tip