Learn More
Ansell MICROFLEX™ MidKnight™ Black MK-296 Nitrile Gloves
SCI_ED
Catalog No. S28991 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S28991 2X-Large
S28986 X-Small
S28987 Small
S28988 Medium
S28989 Large
S28990 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S28991 Supplier Ansell Supplier No. MK296XXL/EMD
This item is not returnable. View return policy
Powder free, nitrile examination gloves with distinct black color hides stains and provides contrast for improved visibility.
  • Distinctive black color conceals dirt, stains and oils
  • Nitrile gloves provide enhanced tactile sensitivity
  • Fully textured gloves provide a secure grip in wet and dry conditions
  • Powder free, ambidextrous, non-sterile, single-use
  • Length: 9.6 in. (24 cm)
  • Tested against both fentanyl and gastric acid
  • Applicable industries include healthcare, life sciences, automotive and machinery/equipment
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 2X-Large
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
Model MK-296
Tensile Strength 27 MPa
Length (Metric) 24.5 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.