Learn More
Medchemexpress LLC HY-107663A 25mg Medchemexpress, MIF-1 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107663A25MG
Encompass_Preferred
Medchemexpress, HY-107663A 25mg MIF-1 (TFA) CAS: MIF-1 TFA, an antibiotic with melanin synthesis inhibitory activity, strongly inhibits melanin formation in Streptomyces bikiniensis NRRLB-1049 and B16 melanoma cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0718
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.