Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Millennia™ 2.0 Adhesion Microscope Slides
GSA_VASDP

Catalog No. STL318B
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
White
Yellow
Sterility:
Non-sterile
Sterile
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility
STL318B Blue Non-sterile
STL318 White Sterile
STL318A Aqua Non-sterile
STL318G Green Non-sterile
STL318L Lilac Non-sterile
STL318Y Yellow Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. STL318B Supplier StatLab™ Supplier No. 318B
This item is not returnable. View return policy

Strong adhesive slides with a hydrophobic surface designed for immunohistochemistry to tolerate harsh chemicals without background staining, tissue artifact or specimen loss.

Specifically designed for IHC, the hydrophobic surfaces of these slides provide immediate specimen tissue adhesion.

  • Features a higher density of binding sites to significantly increase adhesive forces between slide and anchored tissue section
  • Able to tolerate harsh chemicals and reagents without background staining, tissue artifact or specimen loss
  • Compatible with inkjet printers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Length (Metric) 75 mm
Width (Metric) 25 mm
Edges 90°, Ground
Type Adhesion Slide
Frosted 1 End, 1 Side
Thickness (Metric) 1 mm
No. per Pack 144 Pk.
No. per Case 1440 Cs.
Sterility Non-sterile
Material Soda Lime Float Glass
Precleaned Yes
Color Blue
Corner Style 90°
For Use With (Application) IHC
Writing Surface 20 x 25 mm
Dimensions (L x W) 75 x 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.