Learn More
Honeywell Safety Products™ Miller™ SkyORB™ Overhead Rotational Boom Anchor
Catalog No. 19034301 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
SkyORB Anchorage System
SkyORB Shock absorber cable assembly replacement kit
Includes:
Cable assembly kit, base, mast and boom
Cable with swaged ends, two (2) shock absorbers, a clevis and pin, and one (1)carabiner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-034-301 SkyORB Shock absorber cable assembly replacement kit Cable with swaged ends, two (2) shock absorbers, a clevis and pin, and one (1)carabiner
19-034-300 SkyORB Anchorage System Cable assembly kit, base, mast and boom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Any vehicle with a single or tandem axle weight greater than 10,000 lb. (4535kg) or counterweights (not included) in place
Catalog No. 19-034-301 Supplier Honeywell Safety Products™ Supplier No. SORBCABLEKIT
  • $3,105.62
    $2,287.65 / Each of 1
    Save $817.97
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Any vehicle with a single or tandem axle weight greater than 10,000 lb. (4535kg) or counterweights (not included) in place
Pre-engineered boom anchor system that replaces the need for very expensive, custom engineered, fixed overhead anchor systems
Boom anchor provides a safe overhead anchorage that is designed specifically for use around vehicles, utilizing the vehicle weight to secure the anchor for proper fall arrest, eliminating the need for heavy and bulky counter weights
  • Patented energy absorbing design limits forces to the system
  • Includes cable with swaged ends, two (2) shock absorbers, a clevis and pin, and one (1) carabiner
Specifications
Material Stainless Steel
For Use With (Equipment) Any vehicle with a single or tandem axle weight greater than 10,000 lb. (4535kg) or counterweights (not included) in place
Product Type SkyORB Shock absorber cable assembly replacement kit
For Use With (Application) Vehicle loading /unloading and maintenance, where overhead structures are not available for tie-off
Weight (Metric) 3.63kg
Includes Cable with swaged ends, two (2) shock absorbers, a clevis and pin, and one (1)carabiner
Weight (English) 8 lb.
Certifications/Compliance Meets OSHA 1926.502 Subpart M