Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Component for QuickPick™ Rescue Kit: Carabiner Clip

Catalog No. 19826909 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Weight (Metric):
0.04kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
19-826-909 0.04kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-826-909 Supplier Honeywell™ Supplier No. QPSC
  • $342.00
    $250.65 / Each of 1

    Save $91.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Component for QuickPick Rescue Kit

QuickPick Rescue Kits deliver peace of mind during unexpected, stressful peer-rescue situations. The rescuer can remotely attach the system to the suspended worker while remaining securely anchored on the walking/working surface.

  • Quickly raise or lower a fallen worker to safety - avoid suspension trauma
  • Professional rescue solution - enhances worker safety and corporate rescue plan
  • Weight: 0.04kg (0.1 lb.)

Specifications

Product Type Carabiner
Weight (English) 0.1 lb.
For Use With (Equipment) Rescue Kit
Certifications/Compliance OSHA 1926, 1910.66, ANSI A10.32, ANSI Z359.4
Weight (Metric) 0.04kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.