Learn More
Honeywell™ Miller™ Rebar Positioning Assembly
Catalog No. 19134677 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Includes:
Swivel attachment and length adjuster
Catalog No. Includes
19-134-677 Swivel attachment and length adjuster
1 options
Catalog No. 19-134-677 Supplier Honeywell™ Supplier No. 6756RSZ7/
  • $442.00
    $323.65 / Each of 1
    Save $118.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The Miller AirCore™ Tower Climbing harness provides the ultimate in performance, comfort and durability. The Harness Accessory is a rebar chain assembly with locking rebar hook, attached swivel, and two locking snap hooks.
This new line of AirCore harnesses keeps your workers comfortable and safe – increasing productivity. It is designed for the most challenging climbing environments, and is ideal for workers climbing cell phone, TV, radio and utility towers, while building/decommissioning towers, changing out transmission lines/antennas, or installing/ replacing lights.
  • Up to 21% lighter than similar competitive harnesses for reduced fatigue
  • Up to 16% less surface area than other padding keeps workers cooler
  • Removable Belt with tool loops stays in position when disconnected, even with tools attached
  • Improved Cam Buckle is lghtweight, low profile and easy to adjust
  • Rated to 400 lbs. (181.4kg) capacity
  • Meets all applicable OSHA and ANSI standards
Specifications
Includes Swivel attachment and length adjuster
Certifications/Compliance ANSI Z359 compliant
Description Rebar chain assembly with two locking snap hooks (0.75 in. gate opening) and one locking rebar hook (2.5 in. gate opening)