Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ DuraFlex™ 650 Series Full-Body Stretchable Harnesses

Catalog No. 19274710
Change view
Click to view available options
Includes:
Quick connect buckle chest strap and legs straps; Comfort D-pad
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
19-274-710 Quick connect buckle chest strap and legs straps; Comfort D-pad Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-274-710 Supplier Honeywell™ Supplier No. E650QC/UGN

missing translation for 'validMainframeMsg'

Feel the difference with specially-formulated elastomer (stretchable) webbing

With comfort unsurpassed, DuraFlex Harnesses offer wider end-user acceptance. And, a comfortable harness is worn properly, enhancing compliance and safety on the job site.

  • Maximum Comfort: harnesses conform to the shape of the worker, stretching with every movement, so there is no bunching, binding or kinking
  • Increased Productivity: because DuraFlex Harnesses are stretchable, the worker has greater mobility with less fatigue
  • Specially-formulated elastomer webbing stretches to provide greater comfort, improved safety and increased productivity
  • Soft and flexible D-ring back pad further enhances comfort
  • Self-contained label pack encapsulates labels, minimizing damage and loss
  • Integrated and oversized pull-free lanyard rings
  • Hazards: confined Space, fall from height
  • Rated to 400 lb. capacity

Duraflex Harnesses

  • 17-987-605 (Model E650-4/S/MGN): Friction buckle shoulder straps; tongue buckle legs straps; Small/Medium
  • 19-046-150 (Model E650D-4/UBL): Friction buckle shoulder straps; tongue buckle legs straps; Blue; Universal

Duraflex Ultra Harnesses

  • 19-274-710 (Model E650QC/UGN): Quick connect buckle chest strap and legs straps; Comfort D-pad; Universal
  • 18-825-104 (Model E650QC-4/UGN): Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad; Universal
  • 18-825-105 (Model E650QC-4/S/MGN): Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad; Small/Medium

Applications:
Construction, Manufacturing, Wind Energy

Compliance

Meets all applicable OSHA, ANSI and CSA standards

Warranty and Services

Limited lifetime, in the unlikely event that you should discover defects in either workmanship or materials. Does not apply to normal wear and tear or abusive treatment of the product.

TRUSTED_SUSTAINABILITY

Specifications

Color Green, Black
D-Ring Location Back
Size Universal
Product Type DuraFlex 650 Series Full-Body Stretchable Harness
Includes Quick connect buckle chest strap and legs straps; Comfort D-pad
Weight (English) 400 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.