Learn More
Honeywell™ Miller™ DuraFlex™ Harnesses with Tongue-Buckle Leg Straps
Catalog No. 19002910
Change view
Click to view available options
Tongue buckle legs straps
Catalog No.
19-002-910 Tongue buckle legs straps
1 options
Catalog No. 19-002-910 Supplier Honeywell™ Supplier No. E6504/UGN
Ideal for workers who must carry tools and prefer the easy donning and doffing of tongue buckles
  • Stretchable elastic webbing in shoulder and body straps for greater comfort, improved safety, and increased productivity
  • Leg straps of nylon webbing have grommets for strength and durability
  • Feature pull-up-design shoulder adjustment; belt loops to accommodate tool belts, back pads, and accessories; and lanyard clip
  • 1.75 in. (4.4cm) back strap and adjustable chest strap keep shoulder straps from slipping
  • Striped upper webbing visually contrasts with solid-colored leg straps to help the wearer don the harness correctly and with ease
  • Bright design is readily visible from a distance
  • Injection-molded Santoprene™ back D-ring pad is soft and flexible to enhance comfort
  • Sub-pelvic strap better distributes the pressure from the shoulders and legs
  • Back strap also keeps back D-ring in proper position
  • Includes space for inspection tag and personal identification
  • Green
  • Universal size
Order Info
Cat. No. 19-034-402 has side D-rings for worker positioning.
Compliance
Meet all applicable OSHA, ANSI™, and CSA requirements, including ANSI Z359.1-2007.
TRUSTED_SUSTAINABILITY
Specifications
Material Elastomer Webbing
Size Universal
Certifications/Compliance Meets all applicable OSHA, ANSI, and CSA requirements, including ANSIFor Use With (Application) Z359.1-2007.