Learn More
Honeywell™ Miller™ DuraFlex™ Utility Harnesses
Description
With comfort unsurpassed, workers prefer DuraFlex Harnesses, improving safety and productivity. A comfortable harness is more likely to be worn properly, enhancing compliance on the jobsite. And because DuraFlex harnesses are stretchable, the worker has greater mobility with less fatigue.
- Specially-formulated elastomer webbing that stretches to provide greater comfort, improved safety and increased productivity
- Soft and flexible D-ring back pad further enhances comfort
- Self-contained label pack encapsulates labels minimizing damage and loss
- Integrated and oversized pull-free lanyard rings
- Meets all applicable OSHA, ANSI and CSA standards
Specifications
Specifications
| Size | Small/Medium |