Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Equipment Bags For Tools or Tripods

Catalog No. 17987508
Change view
Click to view available options
For hoist, winch or SRL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-987-508 For hoist, winch or SRL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-508 Supplier Honeywell™ Supplier No. 8280H/YL

Carrying bag for hoist, winch or self-retracting lifeline (SRL)

Color: Yellow and Black

TRUSTED_SUSTAINABILITY

Specifications

Product Type Carrying bag
For Use With (Equipment) Hoist, winch, self retracting lifeline
Certifications/Compliance OSHA, ANSI, CSA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.