Learn More
Honeywell Safety Products™ Miller™ HP™ Fall Arrest and Retrieval Harnesses
Catalog No. 191202746
Change view
Click to view available options
Buckle Style:
Qwik-Fit
D-Ring Location:
Back
Size:
2X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style D-Ring Location Size
19-120-2746 Qwik-Fit Back 2X-Large
19-120-2745 Qwik-Fit Back Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-120-2746 Supplier Honeywell Safety Products™ Supplier No. 850T4/XXLGK
Webbing coated with PTFE to extend service life
Superior water and oil repellency and protection against dirt, grime, spills and stains.
  • With lanyard ring, sub-pelvic strap, back strap and back or side D-rings
  • Bright green for high visibility and ease of inspection
  • Friction buckles on the shoulder straps
(This style is also available with front D-ring, side D-rings and mating buckles on the shoulder straps. See below.) Models 850T and 850T-7 have mating buckles on both the leg and chest straps. Models 850T-4 and 850T-58 have tongue buckles on the leg straps and mating buckles on the chest strap.
Compliance
Meet all applicable OSHA, ANSI, and CSA requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE-coated Nylon Webbing
D-Ring Location Back
Certifications/Compliance Meets all applicable OSHA, ANSI, and CSA requirements.
Size 2X-Large
Buckle Style Qwik-Fit