Learn More
Honeywell™ Miller™ Lined Nylon Body Belt
Catalog No. 19074572
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-074-572 Medium
19-074-573 Large
19-074-571 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-074-572 Supplier Honeywell™ Supplier No. 123NL/MBK
Ideal positioning accessory for full-body harness
Made of durable lined nylon; features a tongue buckle and a single D-ring. Meets OSHA, ANSI and CSA Z259.1-95 requirements. A body belt should be used only for work positioning, or in conjunction with a full-body harness.
Note: OSHA requires that full-body harnesses be used for fall protection in the construction industry.
TRUSTED_SUSTAINABILITY
Specifications
D-Ring Location One
Size Medium
Certifications/Compliance Meets OSHA, ANSI and CSA Z259.1-95 requirements.
Length (English) 36 to 44 in.