Learn More
Honeywell™ Miller™ Lined Nylon Body Belt
Supplier: Honeywell™ 123NL/LBK
Description
Ideal positioning accessory for full-body harness
Made of durable lined nylon; features a tongue buckle and a single D-ring. Meets OSHA, ANSI and CSA Z259.1-95 requirements. A body belt should be used only for work positioning, or in conjunction with a full-body harness.
Note: OSHA requires that full-body harnesses be used for fall protection in the construction industry.
Specifications
Meets OSHA, ANSI and CSA Z259.1-95 requirements. | |
One | |
Large | |
42 to 48 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.