Learn More
Honeywell™ Miller™ Manyard HP™ Lanyards with SofStop™ Shock-Absorber
Catalog No. 19034893 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Connector Types:
Locking Snap Hook and D-ring
Catalog No. Connector Types
19-034-893 Locking Snap Hook and D-ring
1 options
Catalog No. 19-034-893 Supplier Honeywell™ Supplier No. 928LSZ7/18INBK
  • $229.51
    $212.65 / Each of 1
    Save $16.86
    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The versatile Miller SofStop Shock Absorber pack is an integral component of various Miller brand web shock-absorbing lanyards. ANSI Z359-2007 Compliant SofStop Shock Absorber Pack has 18 in. length, 1 locking snap hook and 1 D-ring, black.
SofStop Shock Absorber pack is designed with a specially-woven inner core that smoothly expands up to 42 inches (1.1m) to reduce fall arrest forces.
Specifications
Leg Configuration Single-Leg
Connector Types Locking Snap Hook and D-ring