Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Manyard™ HP™ Shock-Absorbing Lanyards

Reduce the force of a fall by up to 80%

$119.00 - $155.50

Specifications

Length (English) 6 ft.
View More Specs

Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19-085-608
View Documents
Honeywell™
216WLSZ7/6FTYL
Yellow Each for $119.00
Add to cart
 
19-169-925
View Documents
Honeywell™
216TWLSZ7/6FTGNC
Hi-Vis Green Each for $155.50
Add to cart
 
Description

Description

  • Specially woven shock-absorbing inner core smoothly expands up to 42 in. (1.1 m) to reduce fall arrest
  • Heavy-duty tubular outer jacket serves as a back-up web lanyard
  • Warning flag provides visual proof unit has been involved in a fall and must be removed from service
  • Locking snap-hook harness connections
  • Single-leg style
  • Hi-vis green

Order Info

Length: 6 ft. (1.8 m)

Compliance

Meets OSHA 1910.66 and OSHA 1926.502; ANSI™ Z359.13 and ANSI A10.32 requirements.

Specifications

Specifications

6 ft.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit