Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Non-Stretch Harnesses with Side D-Rings

Catalog No. 19034818C
Change view
Click to view available options
With side D-rings; Size: X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-034-818C With side D-rings; Size: X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-034-818C Supplier Honeywell™ Supplier No. 80957/XLYK

missing translation for 'validMainframeMsg'

Ideal for use at job sites such as tanks and manholes, where worker must be suspended, raised, or lowered

  • Redistribute pressure from shoulders and legs while providing superior support for buttocks
  • Durable, strong polyester webbing
  • Built-in tongue-buckle waist belt with grommets makes harness easy to don and adds stability when wearer is carrying tools
  • With side D-rings for positioning applications
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Webbing Material 1.75 in. Polyester
Style Non-stretch full body
Size X-Large
Includes Pull Free Lanyard Ring
D-Ring Location Side
Product Type Full-body non-stretch harness
Buckle Style (Leg) Friction Leg Straps
Buckle Style (Shoulder) Friction Shoulder Strap
Certifications/Compliance ANSI A10.14, Z359.1, ASTM 50 hr., CSA Z259.2.2 Type 1, OSHA 1926.502, 1910.66
For Use With (Application) General industry
Capacity (English) 310 lb.
Buckle Style (Chest) Mating Chest Strap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.