Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Non-Stretch Harnesses with Tongue-Buckle Leg Straps

Catalog No. 19069993C
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-06-9993C X-Large
19-069-993A Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-06-9993C Supplier Honeywell™ Supplier No. 80954/XLYK

missing translation for 'validMainframeMsg'

  • Redistribute pressure from shoulders and legs while providing superior support for buttocks
  • Durable, strong polyester webbing
  • Built-in tongue-buckle waist belt with grommets makes harness easy to don and adds stability when wearer is carrying tools
  • Feature tongue-buckle leg straps for quick donning
TRUSTED_SUSTAINABILITY

Specifications

Color Black, Yellow
Webbing Material 1.75 in. Polyester
Buckle Style (Shoulder) Friction Shoulder Strap
Style Subpelvic full body
Certifications/Compliance ANSI A10.32, ANSI Z359.1, OSHA 1926.502
For Use With (Application) Ideal for being suspended, raised or lowered, such as in tanks, manholes and pipe
Buckle Style (Leg) Tongue-buckle Leg Strap
Product Type Subpelvic non-stretch full-body harness
Buckle Style (Chest) Mating Chest Strap
Size X-Large
Capacity (English) 400 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.