Learn More
Honeywell™ Miller™ Non-Stretch Harnesses
Catalog No. 19002990
Change view
Click to view available options
D-Ring Location:
Sliding Back, Front
Catalog No. D-Ring Location
19-002-990 Sliding Back, Front
1 options
Catalog No. 19-002-990 Supplier Honeywell™ Supplier No. 552/UYK
Light and durable full-body harnesses
  • Polyester webbing
  • Back, front and side D-rings
  • Mating buckles on shoulder and leg straps
  • Sub-pelvic strap
  • Pull-free lanyard rings
  • Color: Yellow
  • Size: Universal
TRUSTED_SUSTAINABILITY
Specifications
Material Polyester
Size Universal
D-Ring Location Sliding Back, Front