Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Respirator Bag Belt

Catalog No. 19020612 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-020612 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-020612 Supplier Honeywell™ Supplier No. 6819/ALLBK
  • $27.81
    $24.65 / Each of 1

    Save $3.16

    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Miller Respirator Bag

Accessory for the Miller Respirator Bag

  • Positive snap buckle with friction slide waist adjustment
  • Ideal for workers who wear coveralls (or protective clothing) without a belt
  • One size fits most
  • Quick and easy to use
  • Color: Black

Specifications

Product Type Respirator Bag Belt
Color Black
Size Universal
For Use With (Application) Miller respirator bag
For Use With (Equipment) Miller respirator bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.