Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Respirator Bags

Catalog No. 19020614 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Closure Type:
Hook-and-Loop
Snap
Includes:
Snap loops to attach to a belt, compact half mask
Snap loops to attach to a belt, full face
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Includes
19-020614 Hook-and-Loop Snap loops to attach to a belt, compact half mask
19-020615 Snap Snap loops to attach to a belt, full face
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-020614 Supplier Honeywell™ Supplier No. 6833HS/YL
  • $79.92
    $70.65 / Each of 1

    Save $9.27

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bags provide durable hygienic protection for storing respirators between use

  • Constructed of Millerhyde, a tough open weave nylon that is double laminated with vinyl
  • Lightweight, yet durable
  • Easily cleaned
  • Each bag is equipped with convenient snap loops to attach to a belt
  • Available in a variety of closures
  • Color: Yellow with black straps

Specifications

Product Type Respirator Bag
Closure Type Hook-and-Loop
Color Yellow with black straps
Material Open-weave Nylon, Double-laminated with Vinyl
Includes Snap loops to attach to a belt, compact half mask
For Use With (Application) Durable hygienic protection for storing respirator between use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.