Learn More
Honeywell™ Miller™ Revolution™ Construction Harnesses
Catalog No. 17987604
Change view
Click to view available options
D-Ring Location:
None
Side
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. D-Ring Location
17-987-604 Side
17-987-600 None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-604 Supplier Honeywell™ Supplier No. R10CNTBBDP/S/MGN
Features Type 10 webbing, almost twice as strong as standards require to provide the longest service life possible
  • Size Small/Medium
  • Available with or without removable belt
  • Tongue buckle legs
TRUSTED_SUSTAINABILITY
Specifications
D-Ring Location Side
Size Small/Medium