Learn More
Honeywell™ Miller™ StretchStop™ Lanyard with SofStop Shock Absorber
Catalog No. 19066225 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Connector Types:
2 Locking snap hooks
Leg Configuration:
Single-Leg
Catalog No. Connector Types Leg Configuration
19-066-225 2 Locking snap hooks Single-Leg
1 options
Catalog No. 19-066-225 Supplier Honeywell™ Supplier No. 913SSZ7/6FTGN
  • $424.00
    $390.65 / Each of 1
    Save $33.35
    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Combination of integral SofStop Shock Absorber with stretchable lanyard design
Choose from three options:
  • 6-ft. single leg, 2 locking snap hooks
  • ANSI Z359 Compliant, Two legs, locking snap hook and 2 locking rebar hooks (2.5 in.)
  • ANSI Z359 Compliant, 6-ft. single leg, 1 locking snap hook and 1 locking rebar hook (2.5 in.)
Compliance
ANSI Z359 Compliant.
Specifications
Color Green
Leg Configuration Single-Leg
Length (English) 6 ft.
Connector Types 2 Locking snap hooks