Learn More
Honeywell™ Miller™ Titan™ Fall Protection Kits
Catalog No. 19041451
Change view
Click to view available options
Fall Prot Kit Arl Lft W/bag
Catalog No.
19-041-451 Fall Prot Kit Arl Lft W/bag
1 options
Catalog No. 19-041-451 Supplier Honeywell™ Supplier No. TK4051B/U/6FTAK
Kits include full-body harness, shock-absorbing lanyard, and accessories
These kits include everything needed to operate safely in applications where a dangerous fall is a possibility.
Construction and General Maintenance Fall Protection Kits.
Contain one universal-size Titan full-body, nonstretch harness with mating or tongue buckle leg straps and optional side D-rings, one 6 ft. (1.8m) tubular shock-absorbing lanyard and one 6 ft. (1.8m) cross-arm strap.
Roofing Fall Protection Kit.
Contains one universal-size full-body harness with mating buckle leg straps, one 3 ft. (0.9m) pack-type shock-absorbing lanyard, one reusable roof anchor with attachment D-ring, 5/8 in. (16mm) trailing rope grab, and one 25 ft. (7.6m) rope lifeline.
Aerial Lift Fall Protection Kit.
Contains one universal-size full-body harness with mating buckle leg straps, one attached 6 ft. (1.8m) tubular shock-absorbing lanyard, and a water-resistant storage bag with rail attachment loops.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance OSHA, ANSI
Includes Mating buckle legs, chest and shoulder straps, sub-pelvic support 6 ft. tubular shock-absorbing lanyard, locking snap hook, water resistant storage bag
Product Type Titan aerial lift fall protection kit