Learn More
Honeywell™ Miller™ TranScendor Cable Sleeves
Catalog No. 19074570
Change view
Click to view available options
Includes swivel and carabiner
Catalog No.
19-074-570 Includes swivel and carabiner
1 options
Catalog No. 19-074-570 Supplier Honeywell™ Supplier No. TRCSSC/
Manual pass-through cable sleeve with swivel and carabiner for Vi-Go Ladder Climbing Safety Systems. Fall arrestor cable sleeves are designed to follow the user along the lifeline while ascending or descending, instantly locking in the event of a fall.
Vi-Go Ladder Climbing Safety Systems provide the ultimate in safety with continuous fall protection when climbing fixed ladders. Systems are available in easy-to-install kits or as a build your own option.
  • Uniquely-designed Vi-Go Cable Sleeve locks instantly in the event of a fall
  • Requires manual removal/insertion of cable with intermediate guides
  • The Vi-Go Cable Sleeve is constructed with durable, corrosion-resistant aluminum and stainless steel components
  • Cable sleeve integrated shock-absorbing element provides a dual shock-absorbing system to further reduce fall forces and protects the system and ladder from damage
TRUSTED_SUSTAINABILITY
Specifications
Material Steel
Includes Swivel, carabiner
Strength 5000 lbf.
For Use With (Equipment) Transcendor cable climbing system, harness with front D-ring
Product Type Cable Sleeve
Certifications/Compliance OSHA, ANSI, CSA
Length (Metric) 22.8 cm
Length (English) 9 in.