Learn More
Honeywell™ Miller™ Vi-Go Manual Pass-Through Cable Sleeves
Catalog No. p-4013696
Change view
Click to view available options
Includes:
Manual pass-through
Manual pass-through with carabiner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-074-568 Manual pass-through with carabiner
19-074-567 Manual pass-through
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-074-568 Supplier Honeywell™ Supplier No. TRCSC/
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Vi-Go Ladder Climbing Safety Systems provide the ultimate in safety with continuous fall protection when climbing fixed ladders. Manual Fall Arrestor Cable Sleeves are designed to follow the user along the lifeline while ascending or descending, instantly locking in the event of a fall.
  • Uninterrupted fall protection: Uniquely-designed, patent-pending Vi-Go Cable Sleeve automatically bypasses intermediate cable guides, keeping both hands free for climbing; travels smoothly along cable and locks instantly in the event of a fall.
  • Easy, one-hand operation for attachment/detachment from the system. No pins/chains or external parts to lose or damage.
  • Automatic pass-through cable guides secure lifeline to prevent cable wear and enable the system to accommodate curves.
  • Withstand harsh environments: The Vi-Go Cable Sleeve is constructed with durable, corrosion-resistant aluminum and stainless steel components.
  • Attachment mechanism engineered to prevent incorrect installation, ensuring greater safety. Unit will not open if attached incorrectly.
  • Cable sleeve integrated shock-absorbing element: Provides a dual shock-absorbing system to further reduce fall forces. Protects the system and ladder from damage. Once a fall occurs, the deployed shock absorber indicates the unit must be taken out of service.
  • Accommodate 5/16-inch (8mm) or 3/8-inch (10mm) cable.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cable Sleeve
Includes Manual pass-through with carabiner