Learn More
MilliporeSigma™ Milli-Q™ 7003-7005 Purification Cartridge Kit
Catalog No. IX700XPKT1 Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
IX 7003-7005 Purification Cartridge Kit
IX H 7003-7005 Purification Cartridge Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
IX700XPKT1 IX 7003-7005 Purification Cartridge Kit
IX700XPKT1H IX H 7003-7005 Purification Cartridge Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. IX700XPKT1 Supplier MilliporeSigma™ Supplier No. IX700XPKT1
For full confidence that water is not influencing your analyses, the Milli-Q™ IX pure water system provides a reliable source of high-quality Type 2 pure water to feed your lab’s needs: from general laboratory uses, such as instrument feed or buffer preparation, to your critical applications, such as microbiology culture media or staining solutions
Water Purification Consumables Kit, compatible with the Milli-Q™ IX 7003 and Milli-Q™ IX 7005 systems and includes:
  • IPAK GARD™ H 3-5 Pretreatment Pack (hard water)
  • Activated Carbon for protection against chlorine, organics, bacteria
  • Polyphosphate for protection against scaling and precipitates
  • Depth Filter for protection against particulates and colloids
  • Tank Vent Filter
  • Activated Carbon for volatile organics removal
  • Soda Lime for CO2 removal
  • Membrane Filter to stop particulates and bacteria
TRUSTED_SUSTAINABILITY
Specifications
Capacity 25 L, 50 L and/or 100 L
Flow Rate 3 L/hr. (Milli-Q™ IX 7003), 5 L/hr. (Milli-Q™ IX 7005), 10 L/hr. (Milli-Q™ IX 7010), 15 L/hr. (Milli-Q™ IX 7015)
For Use With (Equipment) Milli-Q™ IX 7003/05/10/15 Water Purification System
Type IX 7003-7005 Purification Cartridge Kit