Learn More
MilliporeSigma MILLIFLEX OASIS 100ML FUNNELS,
Supplier:  MilliporeSigma MMHVWP124
Encompass
Milliflex Oasis 100ml funnels, 0.45µm white plain PVDF Durapore, 24 units per pac
Catalog No. NC1835541