Learn More
GLOBAL FIA INC MILLIGAT HF PUMP W/ CERAMIC
SDP
Supplier:  GLOBAL FIA INC MG2CERXT
Encompass
milliGAT HF pump with Ceramic rotor, PTFEstator, stepper motor, controller,communication cable, and 24VDC powersupply
Catalog No. NC1484839
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.