Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millipak™ Sterile Filters for Milli-Q™ IQ 7000 Ultrapure Water System
GSA_VA

Catalog No. MPGP002A1 Shop All MilliporeSigma Products
Change view
Click to view available options
Description:
 
Gamma Gold Capsule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
MPGP002A1  
MPGPG02A1 Gamma Gold Capsule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPGP002A1 Supplier MilliporeSigma™ Supplier No. MPGP002A1
Add to Cart
Add to Cart

Purification cartridges

  • 0.22μm membrane filter for particulate-free and bacteria-free water at the point of dispense for the Milli-Q IQ 7000 system
  • Sterile unit
  • Delivered hermetically sealed.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description  
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Type Filter, Sterile
Pore Size (Metric) 0.22 μm sterile filter
Purity Meter Included No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.