Learn More
MilliporeSigma Opticap XL5 Millipore Express SHC
Supplier: MilliporeSigma KHGES05TT1
Sterile, inlet connection diam. 1.5 in., pore size 0.2 um, pore size 0.5 um, cartridge nominal length 5 in. (12.5 cm).