Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Oil, Heavy, USP, Spectrum™ Chemical
SDP

Catalog No. p-7157869 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-608 1 L Amber Glass Bottle
18-604-609 4 L Amber Glass Bottle
18-604-610 4 x 4 L Amber Glass Bottle
18-604-611 20 L Poly Drum
18-604-612 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-608 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MI5001LT
Only null left
Add to Cart
Add to Cart

MI500, 8012-95-1

Spectrum™ Chemical Mineral Oil, Heavy, USP is a lubricant laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λ⁴-chromen-1-ylium chloride
SMILES *

Specifications

Acidity To pass test
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 L
Viscosity 34.5-150 mm2
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.