CAS RN 8012-95-1
Paraffin, liquid, pure
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Nujol, for IR spectroscopy
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Paraffin Oil Heavy, MilliporeSigma™
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Paraffin Oil Light, MilliporeSigma™
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Paraffin Oil, BAKER™, J.T. Baker™
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Mineral Oil, Heavy, USP, Spectrum™ Chemical
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N IUPAC Name: 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λ4-chromen-1-ylium chloride SMILES: *
Mineral Oil, Contains Vitamin E as a stabilizer in the range of 10 to 100ppm, U.S.P., J.T. Baker™
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Mineral Oil, Heavy, USP, Spectrum™ Chemical
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N IUPAC Name: 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-1λ4-chromen-1-ylium chloride SMILES: *
Paraffin, liquid, technical
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
International Stock Standard Mixture (2 components), Restek™
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Mineral Oil NF White, Macron Fine Chemicals™
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 0.00 MDL Number: MFCD00131611 InChI Key: FFNDMZIBVDSQFI-UHFFFAOYSA-N Synonym: delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride PubChem CID: 68245 ChEBI: CHEBI:38701 IUPAC Name: 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride SMILES: *
Mineral Oil, Medium, Reagents
CAS: 8012-95-1 Molecular Formula: MFCD00131611 Molecular Weight (g/mol): 452.363