Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Oil NF White, Macron Fine Chemicals™

Catalog No. MK63584 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6358-4 4 L Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-6358-4 Supplier Avantor Supplier No. 635810

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *

Specifications

Color White
Packaging Glass
Quantity 4 L
Grade NF
Physical Form Liquid
Chemical Name or Material Mineral Oil, Light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.