Learn More
Research Products International Corp Mini-300 Orbital-Genie, Benchtop Orbital Shaker with Ratcheting Clamps
SDP
Supplier:  Research Products International Corp 400835
Encompass
The Mini-300 Orbital-Genie with Ratcheting Clamps provides the flexibility for using a mix of different size and shape vessels on a single platform. Compact design fits any lab bench and durable housing withstands years of rugged use.
Unit is equipped with two universal cushioned ratcheting clamps and allows for variable shaking speeds up to 300 RPM.
Catalog No. 50-197-6888