Learn More
Research Products International Corp Mini-300 Orbital-Genie, Benchtop Orbital Shaker for Flask Clamps
SDP
Supplier:  Research Products International Corp 400837
Encompass
The Mini-300 Orbital-Genie for use with flask clamps is ideal for applications using various size flasks or beakers. (Clamps are sold separately.) Compact design fits any lab bench and durable housing withstands years of rugged use.
Platform is equipped to accept multiple clamp sizes and unit allows for variable shaking speeds up to 300 RPM.
Catalog No. 50-197-6899