Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Mini Autobin - 3 Position
SDP

Catalog No. 01259191
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-191 VHB Tape
01-259-136 Magnetic Mount
01-259-172 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-191 Supplier Poltex Supplier No. MINIAUTO3T
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Gravity close bin with 3 compartments and a closing door to help prevent dust accumulation. Holds and dispenses bulk PPE, tubes, and supplies with Wall, Tape, or Magnet Mount options.

  • Gravity-close bin with 3 compartments.
  • Closing the door helps prevent dust accumulation.
  • Holds and dispenses bulk PPE, tubes, and supplies.
  • Choose from Wall Mount (screws/anchors included), HB Tape Mount (high-bond tape included), or Magnet Mount (powerful magnets included).
  • Durable and cleanable PETG construction.
  • Consumables not included.

Specifications

Product Type Front-load Dispenser
Color Clear
Dimensions (L x W x H) 5.5 x 12.3 x 6.2 in.
Holds PPE
Material PETG
Mounting VHB Tape
No. of Compartments 3
Product Style Front-loading with Opening
Capacity 3 Compartment
Edges Rounded
For Use With (Application) Bulk PPE Dispenser, Lab Supplies Holder
Includes High Bond Tape for Mounting
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.