Learn More
Cytiva Fluorodyne II DFL membrane in Mini Kleenpak™ capsules, 0.2 µm removal rating, 200 cm² EFA, ¼-½ in. hose barb connections, pre-sterilized by gamma irradiation, box of 3
Supplier:  Cytiva KA02DFLP2S
Encompass
Capsules are suited to both the filtration of low volumes of fluids (50 mL to 2 L) in the laboratory for filterability purposes, or in a GMP production environment where a certified filter product is essential.
Catalog No. NC0447265
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.