Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Mini Flip-Top Vial, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26734 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Media Type:
9 mL letheen broth
9 mL maximum recovery diluent
Buffered Peptone Water Broth
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
S26734 Buffered Peptone Water Broth
S26735 9 mL letheen broth
S26736 9 mL maximum recovery diluent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26734 Supplier 3M™ Supplier No. BPPFV9BPW
Add to Cart
Add to Cart

Leak-resistant plastic cylinder with flip-open lid that seals securely for safe shipping and handling

  • Hinged lid can be opened with one hand, stays open for easy sample additions
  • Decontaminated through gamma irradiation and ready for use with no additional blending required
  • Flat bottom tube stands on its own, no holder required
  • Leak resistant vial helps prevent leakage during shipping
  • 2 year shelf life from date of manufacture provides long-term readiness

Specifications

Appearance Clear
Broth Type Buffered Peptone Water Broth
Cap Type Flip-top
Certifications/Compliance ISO 9001
Content And Storage 20°C to 25°C
Country of Origin United States
Additive Buffer
Detects Microorganisms
Format Vial
Manufacturing Origin United States
Media Type Buffered Peptone Water Broth
Product Type Bacterial Growth Medium
Sterility Sterile
Target Organism Class Bacteria
Test Type Food and Beverage
Fill Volume 9 mL
For Use With (Application) Microorganism Enumeration
Packaging Type Vial, Case
Quantity 100/Cs.
Specimen Type Food Samples, Environmental Samples
Manufacturing Quality ISO 9001
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.