Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Mini Flip-Top Vial, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26736 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Media Type:
9 mL letheen broth
9 mL maximum recovery diluent
Buffered Peptone Water Broth
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
S26736 9 mL maximum recovery diluent
S26734 Buffered Peptone Water Broth
S26735 9 mL letheen broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26736 Supplier 3M™ Supplier No. BPPFV9MRD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leak-resistant plastic cylinder with flip-open lid that seals securely for safe shipping and handling

  • Hinged lid can be opened with one hand, stays open for easy sample additions
  • Decontaminated through gamma irradiation and ready for use with no additional blending required
  • Flat bottom tube stands on its own, no holder required
  • Leak resistant vial helps prevent leakage during shipping
  • 2 year shelf life from date of manufacture provides long-term readiness

Specifications

Cap Type Flip-top
Content And Storage 20°C to 25°C
Detects Microorganisms
Form Solution
Format Vial
Media Type 9 mL maximum recovery diluent
Preparation Method Pre-filled
Product Type Broth
Sterility Sterile
Target Organism Class Bacteria
Fill Volume 9 mL
For Use With (Application) Sample Preparation, Enumeration of Microorganisms
For Use With (Equipment) 3M Petrifilm Plates
Includes Maximum Recovery Diluent
Packaging Type Vial
Quantity 100/Cs.
Shipping Condition Room Temperature
Specimen Type Food, Food Process Environmental Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.